ContactUsLinkComponent
Stirring-heating block with 15 stirring positions, 15 bore holes, # 57.5 mm, depth 48 mm, distance 65 mm, suitable for 100 ml lab-fl asks (e.g. Duran) and for 150 ml beakers incl. control unit. 1 * 1 items
Catalog # 460-0135
Supplier:  2MAG
Stirring-heating block with 15 stirring positions, 15 bore holes, # 57.5 mm, depth 48 mm, distance 65 mm, suitable for 100 ml lab-fl asks (e.g. Duran) and for 150 ml beakers incl. control unit. 1 * 1 items
Catalog # 460-0135
Supplier:  2MAG
Frequently Bought Together