ContactUsLinkComponent
The Solvent Manager stopper set contains the following parts for replacement: 5 float pins, 5 Plastic plugs, 5 small gasket rings 1 * 1 SET
Catalog # METT51109418
Supplier:  Mettler - Toledo
The Solvent Manager stopper set contains the following parts for replacement: 5 float pins, 5 Plastic plugs, 5 small gasket rings 1 * 1 SET
Catalog # METT51109418
Supplier:  Mettler - Toledo
Frequently Bought Together