Order Entry
Belgium
Orders LinkContactUsLinkComponent
Tissue forceps, 1×2 teeth

Tissue forceps, 1×2 teeth

Stainless steel, 140 mm, 1×2 teeth, Curved, 3×2 mm
Catalog # FSCI11029-14
Marketsource
Supplier:   FINE SCIENCE TOOLS
Tissue forceps, 1×2 teeth
Catalog # FSCI11029-14
Supplier:  FINE SCIENCE TOOLS
Marketsource
Tissue forceps, 1×2 teeth
Tissue forceps, 1×2 teeth

Stainless steel, 140 mm, 1×2 teeth, Curved, 3×2 mm

Price
23,50 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
Spécifications
  • Length:
    140 mm
  • Material:
    Stainless steel
  • Seq:
    01
  • Tip style:
    1×2 teeth
  • Tip size:
    3×2 mm
  • Tip shape:
    Curved
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.