Orders LinkContactUsLinkComponent
Overflow volumetric flasks, class B
Overflow volumetric flasks, class B
420 ml
  HECH42115066
Overflow volumetric flasks, class B
  HECH42115066
Overflow volumetric flasks, class B
Overflow volumetric flasks, class B
420 ml
47,00 €
1 * 1 ST
DURAN® borosilicate glass 3.3.
  • Acid-proof and alkali-resistant markings
  • Very good temperature resistance
  • Very good chemical resistance
For water analysis.
  • Capacity:
    420 ml
  • Seq:
    10