Orders LinkContactUsLinkComponent
Overflow volumetric flasks, class B
Overflow volumetric flasks, class B
157 ml
Catalog # HECH42115030
Overflow volumetric flasks, class B
Catalog # HECH42115030
Overflow volumetric flasks, class B
Overflow volumetric flasks, class B
157 ml
Price
27,30 €
Log in for availability
1 * 1 ST
Product Details & Documents
DURAN® borosilicate glass 3.3.
  • Acid-proof and alkali-resistant markings
  • Very good temperature resistance
  • Very good chemical resistance
For water analysis.
Specifications
  • Capacity:
    157 ml
  • Seq:
    05