Write-on labels
Write-on labels, Vinyl, Black on white, 150×100 mm, 10 labels/pack
BRDY230632
: Brady
These write-on labels provide easy identification for calibration, testing, and repair of tools and equipment.
- Labels are used to track calibration, inspection, or parts and/or equipement
Write-on areas allow date, inspector, or other information to be periodically added.
- Colour:Black on white
- Description:Write-on labels
- Dimensions:150×100 mm
- Material:Vinyl
- Seq:20
- Packed:10 labels/pack