ContactUsLinkComponent
Coolsystem perfect cell freezing . Keep samples cold and dry with ice- free CoolBox/CoolRack CFT, which grips vials for one-handed opening and closing. Then place the prepared vials into a CoolCell for alcohol- free freezing in a -80C freezer. Incl 1 * 1 items
Catalog # 479-0947
Supplier:  BIOCISION
Coolsystem perfect cell freezing . Keep samples cold and dry with ice- free CoolBox/CoolRack CFT, which grips vials for one-handed opening and closing. Then place the prepared vials into a CoolCell for alcohol- free freezing in a -80C freezer. Incl 1 * 1 items
Catalog # 479-0947
Supplier:  BIOCISION
Frequently Bought Together