Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

HPLC columns

Analytical, 4,00 mm, 250 mm
  JOURJR-68185NF
HPLC columns
  JOURJR-68185NF
HPLC columns

Analytical, 4,00 mm, 250 mm

 
354,00 €
   
1 * 1 ST
 

These PEEK columns are designed for naro-bore applications with small amount of samples.

  • Biocompatible
  • High pressure applications
  • Packing devices available
 
  • Column Format:
    Analytical
  • Ø int.:
    4,00 mm
  • Length:
    250 mm
  • Seq:
    10
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.