Order Entry
Belgium
Orders LinkContactUsLinkComponent
HPLC columns

HPLC columns

Analytical, 4,00 mm, 150 mm
Catalog # JOURJR-68184NF
Supplier:   VICI JOUR RESEARCH
HPLC columns
Catalog # JOURJR-68184NF
Supplier:  VICI JOUR RESEARCH
HPLC columns

Analytical, 4,00 mm, 150 mm

Price
307,00 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents

These PEEK columns are designed for naro-bore applications with small amount of samples.

  • Biocompatible
  • High pressure applications
  • Packing devices available
Spécifications
  • Column Format:
    Analytical
  • Ø int.:
    4,00 mm
  • Length:
    150 mm
  • Seq:
    09