Order Entry
Belgium
Orders LinkContactUsLinkComponent
HPLC columns

HPLC columns

Analytical, 4,00 mm, 125 mm
Catalog # JOURJR-68183NF
Supplier:   VICI JOUR RESEARCH
HPLC columns
Catalog # JOURJR-68183NF
Supplier:  VICI JOUR RESEARCH
HPLC columns

Analytical, 4,00 mm, 125 mm

Price
307,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

These PEEK columns are designed for naro-bore applications with small amount of samples.

  • Biocompatible
  • High pressure applications
  • Packing devices available
Specifications
  • Column Format:
    Analytical
  • Ø int.:
    4,00 mm
  • Length:
    125 mm
  • Seq:
    08
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.