Order Entry
Belgium
Orders LinkContactUsLinkComponent
Sample lance

Sample lance

Valve lance, Stainless steel V4A, 550 mm, 32 mm
Catalog # 300-0076
Supplier:   BURKLE
Sample lance
Catalog # 300-0076
Supplier:  BURKLE
Sample lance
Sample lance

Valve lance, Stainless steel V4A, 550 mm, 32 mm

Price
2 480,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Samplers for viscous, paste-like and semi-liquid substances.

  • Optimised opening mechanism (can be completely disassembled)
  • Easy to use and to clean
  • Different layers remain intact particularly with paste-like substances

Designed for use in pharmaceutical applications.

 : DIN 51750.3 and 53242.3

Specifications
  • Ø:
    32 mm
  • Description:
    Valve lance
  • Length:
    550 mm
  • Material:
    Stainless steel V4A