maintenance kit for mp/mpc 104e, 105e, 054z, 055z, 095z, anavac 104, biovac 106 diaphragm pump 1 * 1 items
Catalog # ILMV402045
Supplier: Welch by Gardner Denver
maintenance kit for mp/mpc 104e, 105e, 054z, 055z, 095z, anavac 104, biovac 106 diaphragm pump 1 * 1 items
Catalog # ILMV402045
Supplier: Welch by Gardner Denver
Frequently Bought Together
Gas cartridges, Campingaz®
Supplier: CAMPING GAZ
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Accessories for electronic dispensers, Multipette E3/E3x
Supplier: EPPENDORF
Accessories for Research® Plus pipettes
Supplier: EPPENDORF
Ethanol, GPR RECTAPUR®
Supplier: VWR Chemicals
Accessories for pipetus® / pipetus®-standard / pipetus®-junior
Supplier: HIRSCHMANN
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
VWR®, Accessories for Mechanical Pipettes
Supplier: VWR Collection
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Accessories for accu-jet® pro
Supplier: Brand
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
Supplier: Merck
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Supplier: VWR Chemicals
Set vacuum seal VS22 cpl. 1 * 1 items
Supplier: BUCHI
Vacuum seal PTFE / FKM
Supplier: Heidolph Instruments GmbH & Co.KG