Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Racks for cryogenic vials, polycarbonate

1,2/2,0 ml, 81, Green, 133×133×52 mm
Catalog # DELTM-033V
Supplier:   DELTALAB
Racks for cryogenic vials, polycarbonate
Catalog # DELTM-033V
Supplier:  DELTALAB
Racks for cryogenic vials, polycarbonate

1,2/2,0 ml, 81, Green, 133×133×52 mm

Price
42,50 €
Log in for availability
1 * 1 ST
Product Details & Documents

These cryovials storage boxes are made of extra strong polycarbonate, designed to be used at temperatures between −190 and 121 °C. Transparent covers are printed with a series of squares and are numbered from 1 to 81 for better inventory control.

  • Can be immersed in liquid nitrogen
  • Stackable and have air vents
  • Covers are keyed to the base to prevent misalignment

Boxes made to accept 81 tubes have a white base and a coloured grid (in which each well is numbered).

 : Each box is supplied with an innovative vial picker for easy vial removal.

Specifications
  • For tubes:
    1,2/2,0 ml
  • No. of Holes:
    81
  • Colour:
    Green
  • Seq:
    03
  • W×D×H:
    133×133×52 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.