Order Entry
Belgium
Orders LinkContactUsLinkComponent
Ring weights for flasks

Ring weights for flasks

Viny coated lead ring weight, Erlenmeyer 250 - 1000 ml, 51 mm
Catalog # DELT192471
Supplier:   DELTALAB
Ring weights for flasks
Catalog # DELT192471
Supplier:  DELTALAB
Ring weights for flasks

Viny coated lead ring weight, Erlenmeyer 250 - 1000 ml, 51 mm

Price
143,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Closed flask weight rings with heavy vinyl coating, which is ideal to improve the stability of Erlenmeyer flasks in a water bath.

  • High chemical resistance
Specifications
  • Description:
    Viny coated lead ring weight
  • Seq:
    02
  • For:
    Erlenmeyer 250 - 1000 ml
  • Ø int.:
    51 mm
Recommandations

Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".

Dans le cas contraire, vous recevrez des recommandations génériques.