Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Gas sampling balloons

Gas sampling balloon, 200 mm, 4,0 L
Catalog # KART1233
Supplier:   KARTELL
Gas sampling balloons
Catalog # KART1233
Supplier:  KARTELL
Gas sampling balloons
Gas sampling balloons

Gas sampling balloon, 200 mm, 4,0 L

Price
48,80 €
Log in for availability
1 * 1 ST
Product Details & Documents

Gas collection bags made from latex.

Designed for general use in laboratories.

 : Available in different volumes and diameters.

Specifications
  • Ø:
    200 mm
  • Volume:
    4,0 L
  • Description:
    Gas sampling balloon