VWR® Basic Bouffant Caps, with Latex
With latex, White, 530 mm
Supplier: VWR Collection
Aller à :
Product Details & Documents
VWR® Basic bouffant caps are manufactured from a strong, lightweight and breathable, spunbonded polypropylene (SPP) fabric, which protects against dirt, grime, and certain dry particulates, in non hazardous environments.
- Latex
- Lightweight and breathable first line barrier protection
- Tested and manufactured in an ISO certified facility that is lot-controlled and validated through independent laboratory testing
This garment is ideal for less critical areas or pre-gowning entry rooms.
Spécifications
- Ø:530 mm
- Colour:White
- Description:With latex
- Seq:05
- Cat. No.:113-1461
- Garment application:Cleanroom
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.