VWR® Basic Bouffant Caps, with Latex
With latex, White, 610 mm
Fournisseur: VWR Collection
Jump to:
Product Details & Documents
VWR® Basic bouffant caps are manufactured from a strong, lightweight and breathable, spunbonded polypropylene (SPP) fabric, which protects against dirt, grime, and certain dry particulates, in non hazardous environments.
- Latex
- Lightweight and breathable first line barrier protection
- Tested and manufactured in an ISO certified facility that is lot-controlled and validated through independent laboratory testing
This garment is ideal for less critical areas or pre-gowning entry rooms.
Specifications
- Ø:610 mm
- Colour:White
- Description:With latex
- Seq:07
- Cat. No.:113-1462
- Garment application:Cleanroom