Order Entry
Belgium
Orders LinkContactUsLinkComponent
Copper, powder

Copper, powder

Glass bottle for solids, 100 g
Catalog # A16243.22
Flammable
Copper, powder
Catalog # A16243.22
CAS Number:  7440-50-8
Copper, powder
Copper, powder

Glass bottle for solids, 100 g

CAS Number:  7440-50-8
Price
29,50 €
Log in for availability
1 * 100 g
Product Details & Documents
  • CAS Number:7440-50-8
  • Formula:Cu
  • MDL Number:MFCD00010965
  • Molecular Weight:63.55 g/mol
  • Boiling Pt:2567 °C (1013 hPa)
  • Melting Pt:1083.4 °C
  • Density:8.94 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-159-6
  • UN:3089
  • ADR:4.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.