Copper, powder
Glass bottle for solids, 500 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
- CAS Number:7440-50-8
- Formula:Cu
- MDL Number:MFCD00010965
- Molecular Weight:63.55 g/mol
- Boiling Pt:2567 °C (1013 hPa)
- Melting Pt:1083.4 °C
- Density:8.94 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:231-159-6
- UN:3089
- ADR:4.1,III
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g



