VWR® Basic Frocks, SMS
Blue, XL
Supplier: VWR Collection
Jump to:
Product Details & Documents
These SMS frocks are manufactured from a latex-free, multi-layer spunbonded - melt-blown - spunbonded (SMS) fabric, that offers high tensile strength while remaining soft, comfortable and breathable.
- Snap collar, raglan sleeves, snap closure
- Fluid and particulate barrier
- Light chemical splash resistance
Colour: White or blue
Specifications
- Clothing Size:XL
- Colour:Blue
- Seq:09
- Cat. No.:113-1175
- Garment application:Cleanroom|Laboratory
- Gender:Unisex
- Pockets:Without pockets
- Closure type:Press studs / snaps