Orders LinkContactUsLinkComponent
VWR® Basic Cleanroom Overalls, SMS
VWR® Basic Cleanroom Overalls, SMS
Without hood, Blue, M
Catalog # 113-1091
Supplier:   VWR Collection
VWR® Basic Cleanroom Overalls, SMS
Catalog # 113-1091
Supplier:  VWR Collection
VWR® Basic Cleanroom Overalls, SMS
VWR® Basic Cleanroom Overalls, SMS
Without hood, Blue, M
Price
197,00 €
Log in for availability
1 * 25 ST
Product Details & Documents
These cleanroom overalls are manufactured from a latex-free, multi-layer spun-bonded - meltblown - spun-bonded (SMS) fabric, that offers high tensile strength while remaining soft, comfortable and breathable.
  • Tapered collar, inset sleeves, zip closure, elasticated cuffs
  • Fluid and particulate barrier
  • Light chemical splash resistance
Specifications
  • Clothing Size:
    M
  • Colour:
    Blue
  • Description:
    Without hood
  • Seq:
    08
  • Cat. No.:
    113-1091
  • Garment application:
    Cleanroom|Laboratory