Order Entry
Belgium
Orders LinkContactUsLinkComponent
VWR® Basic Cleanroom Lab Coats, SPP, Disposable

VWR® Basic Cleanroom Lab Coats, SPP, Disposable

White, XXL, 100 cm
Catalog # 113-1115
Supplier:   VWR Collection
VWR® Basic Cleanroom Lab Coats, SPP, Disposable
Catalog # 113-1115
Supplier:  VWR Collection
VWR® Basic Cleanroom Lab Coats, SPP, Disposable
VWR® Basic Cleanroom Lab Coats, SPP, Disposable

White, XXL, 100 cm

Price
98,00 €
Log in for availability
1 * 30 ST
Product Details & Documents

VWR® Basic SPP lab coats are manufactured from a strong, lightweight and breathable spunbonded polypropylene (SPP) fabric, which protects against dirt, grime, and certain dry particulates in non hazardous environments.

  • Tapered collar, inset sleeves, snap closure
  • Elasticated or knitted cuffs, no pockets or three pockets
  • Lightweight and breathable first line barrier protection

This garment is ideal for less critical areas or pre-gowning entry rooms. It is tested and manufactured in an ISO certified facility and lot controlled and validated through independent laboratory testing.

Emballage: 25 or 30 coats per carton.

Specifications
  • Clothing Size:
    XXL
  • Colour:
    White
  • Length:
    100 cm
  • Seq:
    24
  • Cat. No.:
    113-1115
  • Garment application:
    Cleanroom|Laboratory|Industrial
  • Gender:
    Unisex
  • Pockets:
    With pockets
  • Closure type:
    Press studs / snaps
  • Weight:
    45 g/m²