Orders LinkContactUsLinkComponent
VWR® Basic Lab Coats, SMS, Disposable
VWR® Basic Lab Coats, SMS, Disposable
White, L, 101,60 cm
Catalog # 113-1133
Supplier:   VWR Collection
VWR® Basic Lab Coats, SMS, Disposable
Catalog # 113-1133
Supplier:  VWR Collection
VWR® Basic Lab Coats, SMS, Disposable
VWR® Basic Lab Coats, SMS, Disposable
White, L, 101,60 cm
Price
193,00 €
Log in for availability
1 * 30 ST
Product Details & Documents
SMS lab coats are manufactured from a latex-free, multi-layer spunbonded-melt-blown-spunbonded (SMS) fabric that offers high tensile strength, while remaining soft, comfortable and breathable.
  • Tapered collar, inset sleeves and snap closure
  • Knitted cuffs and three pockets
  • Fluid and particulate barrier
  • Light chemical splash resistance
Colour: White or blue
Specifications
  • Clothing Size:
    L
  • Colour:
    White
  • Length:
    101,60 cm
  • Cat. No.:
    113-1133
  • Garment application:
    Cleanroom|Laboratory
  • Gender:
    Unisex
  • Pockets:
    With pockets
  • Closure type:
    Press studs / snaps