Orders LinkContactUsLinkComponent
VWR® Basic Cleanroom Overalls, SPP
VWR® Basic Cleanroom Overalls, SPP
Without hood, White, 3XL
Catalog # 113-1108
Supplier:   VWR Collection
VWR® Basic Cleanroom Overalls, SPP
Catalog # 113-1108
Supplier:  VWR Collection
VWR® Basic Cleanroom Overalls, SPP
VWR® Basic Cleanroom Overalls, SPP
Without hood, White, 3XL
Price
122,00 €
Log in for availability
1 * 25 ST
Product Details & Documents
VWR® Basic SPP overalls are manufactured from a strong, lightweight and breathable, spunbonded polypropylene (SPP) fabric, which protects against dirt, grime, and certain dry particulates, in non hazardous environments.
  • Tapered collar, inset sleeves, zip closure, elasticated cuffs
  • Lightweight and breathable first line barrier protection
This garment is ideal for less critical areas or pre-gowning entry rooms.
Tested and manufactured in an ISO certified facility also lot-controlled and validated through independent laboratory testing.
Specifications
  • Clothing Size:
    3XL
  • Colour:
    White
  • Description:
    Without hood
  • Seq:
    05
  • Cat. No.:
    113-1108
  • Garment application:
    Cleanroom|General Purpose|Laboratory