ContactUsLinkComponent
Refrigeration bath circulator steel, sc150-g50, -28 to +150 degree c, 850 w cooling capacity at 20c, 6-12 l bath volume, 2.0 kw hea ter, pump 17 l/min & 300 mbar usb ul/csa, 3 years warranty 230v/50hz 1 * 1 items
Catalog # HAAK153-6501
Supplier:  HAAKE
Refrigeration bath circulator steel, sc150-g50, -28 to +150 degree c, 850 w cooling capacity at 20c, 6-12 l bath volume, 2.0 kw hea ter, pump 17 l/min & 300 mbar usb ul/csa, 3 years warranty 230v/50hz 1 * 1 items
Catalog # HAAK153-6501
Supplier:  HAAKE
Frequently Bought Together