Order Entry
Belgium
Orders LinkContactUsLinkComponent
Redox/ORP electrodes

Redox/ORP electrodes

Platinum ORP (redox) electrode, 0...±2000 mV, Single junction, refillable, S7, Glass, 0...+100 °C, 12×110 mm, General purposes
Catalog # CONSSO60X
Supplier:   CONSORT
Redox/ORP electrodes
Catalog # CONSSO60X
Supplier:  CONSORT
Redox/ORP electrodes
Redox/ORP electrodes

Platinum ORP (redox) electrode, 0...±2000 mV, Single junction, refillable, S7, Glass, 0...+100 °C, 12×110 mm, General purposes

Price
184,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Connection:
    S7
  • Ø×L:
    12×110 mm
  • For:
    General purposes
  • Type:
    Single junction, refillable
  • Description:
    Platinum ORP (redox) electrode, 0...±2000 mV
  • Material:
    Glass
  • Temp.:
    0...+100 °C
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.