Order Entry
Belgium
Orders LinkContactUsLinkComponent
Spinner flasks, disposable, Corning®

Spinner flasks, disposable, Corning®

Flasks with two vented caps on the side arms, 3000 ml, side arm necks thread GL 45, 2, 1000-3000 ml, 188×259 mm
Catalog # 734-2290
Supplier:   Corning
Spinner flasks, disposable, Corning®
Catalog # 734-2290
Supplier:  Corning
Spinner flasks, disposable, Corning®
Spinner flasks, disposable, Corning®

Flasks with two vented caps on the side arms, 3000 ml, side arm necks thread GL 45, 2, 1000-3000 ml, 188×259 mm

Price
1 130,00 €
Log in for availability
1 * 4 ST
Product Details & Documents

The Corning® disposable spinner flask system comes ready-to-use with paddle and integrated magnet, eliminating the need for time-consuming assembly or cleaning and reassembly.

  • Paddle size and height is optimised for each vessel size
  • Integrated magnet provides smooth, even rotation at required speeds on slow-speed stirrers
  • Heat build-up in the vessel is reduced by means of a specially designed flange that raised the vessel off the stir-plate surface for the 125 and 500 ml flasks only

Molded from virgin polystyrene and gamma-irradiated, each spinner flask system assures a clean and sterile unit that is nonpyrogenic. No more concerns with detergent residues or contamination.

Made of USP Class VI and ISO 10993-18 polystyrene, the vessel is comparable to conventional glass spinner flasks for growth of suspension cell lines and any attachment-dependent cultures using microcarrier beads.

Specifications
  • Ø×H:
    188×259 mm
  • Recommended working volume:
    1000-3000 ml
  • Capacity:
    3000 ml
  • Description:
    Flasks with two vented caps on the side arms
  • Seq:
    07
  • Neck:
    side arm necks thread GL 45
  • No. of side arms:
    2
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Modified impeller assembly for magnetically-driven 36 L Corning® Proculture® spinner flask, reusable
    Modified impeller assembly for magnetically-driven 36 L Corning® Proculture® spinner flask, reusable
    CORN401901
    Log in for availability
    1 * 1 ST - 1 340,00 €

Product Information

  • Catalog #
    Thread
    Availability
    Price
  • Vent cap, 0,2 μm vent, for 500, 1000 and 3000 ml spinner flasks
    Vent cap, 0,2 μm vent, for 500, 1000 and 3000 ml spinner flasks
    734-4102
    GL45
    Log in for availability
    1 * 4 ST - 44,90 €

Product Information

  • Catalog #
    Thread
    Availability
    Price
  • Side arm aseptic transfer cap, dip tube with 0,2 μm vent, male luer lock, for 500 ml spinner flask
    Side arm aseptic transfer cap, dip tube with 0,2 μm vent, male luer lock, for 500 ml spinner flask
    734-4101
    GL45
    Log in for availability
    1 * 2 ST - 449,00 €

Product Information

  • Catalog #
    Availability
    Price
  • Flat cap, PE, disposable, orange, for 500, 1000 and 3000 ml spinner flasks
    Flat cap, PE, disposable, orange, for 500, 1000 and 3000 ml spinner flasks
    CORN3568
    Log in for availability
    1 * 4 ST - 41,20 €

Product Information

  • Catalog #
    Thread
    Availability
    Price
  • Side arm aseptic transfer cap, dip tube with 0,2 μm vent, male luer lock, for 3000 ml spinner flask
    Side arm aseptic transfer cap, dip tube with 0,2 μm vent, male luer lock, for 3000 ml spinner flask
    734-4100
    GL45
    Log in for availability
    1 * 2 ST - 449,00 €

Product Information

  • Catalog #
    Thread
    Availability
    Price
  • Side arm aseptic transfer cap, dip tube with 0,2 μm vent, male luer lock, for 1000 ml spinner flask
    Side arm aseptic transfer cap, dip tube with 0,2 μm vent, male luer lock, for 1000 ml spinner flask
    734-4098
    GL45
    Log in for availability
    1 * 2 ST - 449,00 €