Dawson Nbz AM resin (100 - 200 mesh) 1 * 5 g
Catalog # 8.55131.0005
Supplier: Novabiochem (Merck biosciences)
Dawson Nbz AM resin (100 - 200 mesh) 1 * 5 g
Catalog # 8.55131.0005
Supplier: Novabiochem (Merck biosciences)
Frequently Bought Together
Microcentrifuge tubes, 0,5/1,5/2,0 ml
Supplier: Brand
Storage plates, 1536-well, MASTERBLOCK®
Supplier: Greiner Bio-One
N-Fmoc-L-valine
Supplier: Novabiochem (Merck biosciences)
VWR®, Ice Pans, with or without Lid
Supplier: VWR Collection
Tissue vial box, blue, WHEATON® CryoFILE
Supplier: DWK Life Sciences
Reagent/centrifuge tubes, round bottom, with or without push cap
Supplier: SARSTEDT
NANOPLATE 26K 240-WELL 1 * 10 items
Supplier: QIAGEN
Screw cap microcentrifuge tubes, SuperClear®
Supplier: Labcon
VWR®, PCR/qPCR Plates, 96-Wells, Plastic
Supplier: VWR Collection
Cover mats for deep well plates
Supplier: Brand