Screw caps
Screw caps, black, cap size N-15, White styrene/butadiene
Catalog # WITG9.351.103
Supplier: witeg Labortechnik
Jump to:
Product Details & Documents
These black PP screw caps have a white styrene/butadiene seal.
Specifications
- Septum:White styrene/butadiene
- Description:Screw caps, black, cap size N-15