Order Entry
Belgium
Orders LinkContactUsLinkComponent
Brushes for Erlenmeyer flasks

Brushes for Erlenmeyer flasks

Erlenmeyer piston brush, with fallow bristles, Natural bristle, 130 mm, 50 mm, Wire, 340 mm
Catalog # REIT10000401
Brushes for Erlenmeyer flasks
Catalog # REIT10000401
Supplier:  
Brushes for Erlenmeyer flasks

Erlenmeyer piston brush, with fallow bristles, Natural bristle, 130 mm, 50 mm, Wire, 340 mm

Discontinued
Product Details & Documents

Brushes with natural or nylon bristles and wire handle.

Specifications
  • Bristle material:
    Natural bristle
  • Brush head Ø:
    50 mm
  • Handle material:
    Wire
  • Description:
    Erlenmeyer piston brush, with fallow bristles
  • Seq:
    08
  • Total length:
    340 mm
  • Brush head length:
    130 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.