Orders LinkContactUsLinkComponent
Dressing forceps, Cushing type
Dressing forceps, Cushing type
Cushing type, TC Gold (hard metal insert), Straight, blunt, 175 mm
Catalog # MART12-254-17
Supplier:   KLS Martin Group
Dressing forceps, Cushing type
Catalog # MART12-254-17
Supplier:  KLS Martin Group
Dressing forceps, Cushing type
Cushing type, TC Gold (hard metal insert), Straight, blunt, 175 mm
Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
Stainless steel forceps.
  • TC Gold: Tungsten Carbide inlays provide an exceptionally long service life of the instruments
Specifications
  • Type:
    Straight, blunt
  • Description:
    Cushing type, TC Gold (hard metal insert)
  • Length:
    175 mm