BUFFER SOLUTION PH9 1 * 500 mL
FFCHFF1770500
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
: VWR Chemicals
Filtration capsules, AcroPak™
: Cytiva (Formerly Pall Lab)
VWR®, Conductivity standard solutions
: VWR Chemicals
VWR®, Nitrile Gloves
: VWR Collection
BUFFER SOLUTION PH4 1 * 500 mL
: FF CHEMICALS