BUFFER SOLUTION PH7 1 * 500 mL
Catalog # FFCHFF1710500
Supplier: FF CHEMICALS
Frequently Bought Together
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Waterproof digital thermometers, HACCP, testo 108 / 108-2
Supplier: TESTO
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
Detergents and cleaning agents, LABWASH® Premium
Supplier: VWR Chemicals
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Food thermometer, HACCP, testo 106
Supplier: TESTO
VWR®, Centrifuge Tubes, High Performance, Flat or Plug Cap, PP
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
BUFFER SOLUTION PH4 1 * 500 mL
Supplier: FF CHEMICALS
Bottles, Wide Neck with Screw Cap
Supplier: RIXIUS