BUFFER SOLUTION PH2 1 * 500 mL
Catalog # FFCHFF1600500
Supplier: FF CHEMICALS
Frequently Bought Together
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
Acrodisc® One PSF Syringe Filters, wwPTFE Membrane
Supplier: Cytiva (Formerly Pall Lab)
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
VWR®, Nitrile Gloves
Supplier: VWR Collection
Maintenance solutions for electrodes
Supplier: Mettler - Toledo
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
Millex Samplicity® Filters
Supplier: Merck
Hydrochloric acid 5 mol/l (5 N)
Supplier: FF CHEMICALS