Dip slides
Total with Neutr/ Staph, 120
Catalog # LIOF53530
Supplier: LIOFILCHEM
Jump to:
Product Details & Documents
Flexible contact slide for microbiology screening of surfaces.
- ISO 18593
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pk:120
- Description:Total with Neutr/ Staph