Orders LinkContactUsLinkComponent
Dip slides
Dip slides
Total/Enterobac + Neutraliser, 120
Catalog # LIOF53526
Supplier:   LIOFILCHEM
Dip slides
Catalog # LIOF53526
Supplier:  LIOFILCHEM
CAS Number:  
Dip slides
Dip slides
Total/Enterobac + Neutraliser, 120
CAS Number:  
Price
281,00 €
Log in for availability
1 * 120 ST
Product Details & Documents
Flexible contact slide for microbiology screening of surfaces.
  • ISO 18593
Documents
Specifications
Specifications
  • Pk:
    120
  • Description:
    Total/Enterobac + Neutraliser