Orders LinkContactUsLinkComponent
Pipette tips
Pipette tips
With collar, 100 - 1000 µl, -, Blue, Eppendorf, Nichiryo, Biohit, Brand, Gilson, Treff, Menarini, Bulk (10×1000)
Catalog # DELT200012
Supplier:   DELTALAB
Pipette tips
Catalog # DELT200012
Supplier:  DELTALAB
Pipette tips
With collar, 100 - 1000 µl, -, Blue, Eppendorf, Nichiryo, Biohit, Brand, Gilson, Treff, Menarini, Bulk (10×1000)
Price
183,00 €
Log in for availability
1 * 10.000 ST
Product Details & Documents
Specifications
  • Packed:
    Bulk (10×1000)
  • Sterile:
    -
  • Volume:
    100 - 1000 µl
  • Seq:
    15
  • Tip style:
    With collar
  • Tip colour:
    Blue
  • For use with:
    Eppendorf, Nichiryo, Biohit, Brand, Gilson, Treff, Menarini