Order Entry
Belgium
Orders LinkContactUsLinkComponent
Solid phase extraction cartridges, HyperSep™ C18

Solid phase extraction cartridges, HyperSep™ C18

Cartridge, C18, 40 - 60 µm, 75 ml, 10000 mg
Catalog # 549-0693
Solid phase extraction cartridges, HyperSep™ C18
Catalog # 549-0693
Solid phase extraction cartridges, HyperSep™ C18
Solid phase extraction cartridges, HyperSep™ C18

Cartridge, C18, 40 - 60 µm, 75 ml, 10000 mg

Price
274,00 €
Log in for availability
1 * 10 ST
Product Details & Documents

These columns feature a highly retentive alkyl-bonded phase for non polar to moderately polar compounds.

  • Hydrophobic reversed phase
  • Retains most organic analytes from aqueous matrices
Specifications
  • Column Format:
    Cartridge
  • Particle Size:
    40 - 60 µm
  • Capacity:
    75 ml
  • Seq:
    12
  • Bed weight:
    10000 mg
  • Phase:
    C18