Elmasonic X-tra basic 2500 - ultrasonic cleaner Int.Dem. of tank (mm) 750x650x520, Volume: 255 liters, Serv. Cap. 215 1 * 1 items
Catalog # ELMA1027776
Supplier: ELMA SCHMIDBAUER
Elmasonic X-tra basic 2500 - ultrasonic cleaner Int.Dem. of tank (mm) 750x650x520, Volume: 255 liters, Serv. Cap. 215 1 * 1 items
Catalog # ELMA1027776
Supplier: ELMA SCHMIDBAUER
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Sealing film for manual application, Parafilm® M
Supplier: HEATHROW
VWR®, Nitrile Gloves
Supplier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR® Facial Tissues
Supplier: VWR Collection
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Nitrile gloves, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection