Orders LinkContactUsLinkComponent
Accessories for Lambda 25/35
Accessories for Lambda 25/35
Rectangular cell holder with pathlengths from 10 to 100 mm
Catalog # PERCB0080821
Supplier:   PerkinElmer
Accessories for Lambda 25/35
Catalog # PERCB0080821
Supplier:  PerkinElmer
Accessories for Lambda 25/35
Rectangular cell holder with pathlengths from 10 to 100 mm
Price
2 160,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    Rectangular cell holder with pathlengths from 10 to 100 mm