Orders LinkContactUsLinkComponent
Closures for headspace vials
Closures for headspace vials
Blue PTFE coated silicone Bi-metal magnetic cap, pre-assembled
Catalog # PERCN6356566
Supplier:   PerkinElmer
Closures for headspace vials
Catalog # PERCN6356566
Supplier:  PerkinElmer
Closures for headspace vials
Blue PTFE coated silicone Bi-metal magnetic cap, pre-assembled
Price
593,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Caps for 20 ml glass headspace crimp top round bottom vials which have a 23 mm OD and 75,5 mm height.
Specifications
  • Description:
    Blue PTFE coated silicone Bi-metal magnetic cap, pre-assembled
  • Seq:
    18