Order Entry
Belgium
Orders LinkContactUsLinkComponent
Visual colour kits, aluminium, MQuant®, Supelco®

Visual colour kits, aluminium, MQuant®, Supelco®

Aluminium test, RP for 1.18386.0001, Chromazurol S, MCD, 150 Tests
Catalog # 1.18452.0002
Supplier:   Merck
Visual colour kits, aluminium, MQuant®, Supelco®
Catalog # 1.18452.0002
Supplier:  Merck
CAS Number:  
Visual colour kits, aluminium, MQuant®, Supelco®
Visual colour kits, aluminium, MQuant®, Supelco®

Aluminium test, RP for 1.18386.0001, Chromazurol S, MCD, 150 Tests

CAS Number:  
Price
283,00 €
Log in for availability
1 * 1 EA
Product Details & Documents

Quick tests, visual colour comparison.

  • Results in approximately 1 minute
  • Easy to use
  • Application-oriented measuring ranges
  • Portable
  • A range of economical refill packs (RP) are available

MQuant® with colour card comparator

By using cuvettes with a long light path, the MQuant® (MCC) test kits are especially suited for measuring low ion concentrations (some as low as 0,005 mg/L). These test kits are especially appropriate for analysing drinking water, spring water, purified and mineral water, as well as cooling and boiler water.

MQuant® with colour disc comparator MCD

Using the comparator disc, these test kits are especially suitable for the analysis of turbid and coloured waters. The comparator disc is a practical and robust colour comparison device that provides reliable measurements, even in humid environments. Ideal for samples of medium to high concentration in applications ranging from testing bottled water to wastewater and other industrial water testing.

Delivery information: Each test kit contains all the necessary reagents, reaction cuvettes, colour comparison chart or comparator wheel and extensive instructions for use.

Specifications

Specifications

  • Chemical method:
    Chromazurol S
  • Pk:
    150 Tests
  • Description:
    Aluminium test, RP for 1.18386.0001
  • Type:
    MCD

Specifications