Sample cups for analysers
Sample cups for auto sampler, PP, no known solvent at room temperature, 2,5 ml, Translucent
Catalog # PERCB3001566
Supplier: PerkinElmer
Jump to:
Product Details & Documents
Disposable.
These sample cups are of heavy-duty construction to prevent cracking or tipping. Their conical design allows for use of nearly the entire sample. For concentrated acids including HF for analyses.
Specifications
- Capacity:2,5 ml
- Colour:Translucent
- Description:Sample cups for auto sampler, PP, no known solvent at room temperature
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.