Orders LinkContactUsLinkComponent
Closures
Degassing shutters with Gore disc and splash guard, 45 mm, Black, HDPE
Catalog # RIXI8-6023-45
Closures
Catalog # RIXI8-6023-45
Closures
Closures
Degassing shutters with Gore disc and splash guard, 45 mm, Black, HDPE
Price
62,00 €
Connexion Pour la disponibilité
1 * 20 ST
Product Details & Documents
Spécifications
  • Thread:
    45 mm
  • Colour:
    Black
  • Description:
    Degassing shutters with Gore disc and splash guard
  • Material:
    HDPE
  • Seq:
    6