Labels, hazard warning
Hazard labels: warning sign on roll, RL 334-TRI 100-B7541, Polyester, Black on yellow, 100×73 mm, 250 labels/roll
Catalog # BRDY223341
Supplier: Brady
Jump to:
Product Details & Documents
These self adhesive signs are designed to warn people to be careful and to take precautions of potential hazards.
Available in different sizes.
Certifications: Conforms to Health and Safety (Safety Signs and Signals) Regulations 1996.
Specifications
- Colour:Black on yellow
- Description:Hazard labels: warning sign on roll
- Dimensions:100×73 mm
- Material:Polyester
- Seq:18
- Model:RL 334-TRI 100-B7541
- Packed:250 labels/roll
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



