Multi-parameter meter (pH/conductivity/oxygen), handheld, SensoDirect 150
The SensoDirect 150 is designed for multi purpose operation and measures pH/Redox, dissolved oxygen and conductivity/TDS.
- Data logger function, results can be downloaded to a PC for further analysis
- Automatic calibration + buffer recognition
- Large LCD display, intuitive user interface, HOLD function
- Flip out stand for bench top use
- Rugged protective casing for field use and built-in electrode holder for safe storage
Typical applications include: Drinking water, cooling/boiler water, waste water, pool water, surface water, water treatment companies, industrial and governmental laboratories.
Levering: Supplied with batteries, pH electrode, temperature probe, pH buffer set 4,00 / 7,00, instruction manual, guarantee sheet and sturdy case.
The pH / Con set additionally includes a conductivity probe.
The pH / Oxi set additionally includes an oxygen sensor, electrolyte, and membrane heads.
The pH / Con / Oxi set additionally includes a conductivity probe, an oxygen sensor, electrolyte, and membrane heads.
- Description:SensoDirect 150, pH / Con / Oxi set, with pH electrode, oxygen sensor, temperature and conductivity probe
- Seq:01
- DO concentration resolution:0,1 % / 0,1 mg/L
- Conductivity resolution:0,1 µS (0 - 200,0 µS) / 0,001 mS (0,2 - 2,000 mS) / 0,01 mS (2 - 20,00 mS) / 0,1 mS (20 - 200,00 mS)
- DO accuracy:±0,4 mg/l / ± 0,7%
- Data logging capacity:16000 data sets
- Power supply:4×1,5 V AA battery or DC 9 V adapter
- mV resolution:1
- TDS accuracy:± 2 % F.S. + 1 digit
- Conductivity accuracy:± 2 % F.S. + 1 digit
- W×D×H:76×38×203 mm
- Temperature range:0...+60 °C
- mV range:±1999
- Conductivity reference temperature:0 - 60 °C
- pH range:0 - 14,00
- Temperature compensation:Automatic/manual (0...+100 °C)
- pH calibration:3-point calibration; pH 7, pH 4, and pH 10
- Temperature resolution:0,1 °C
- DO concentration:0 - 20,0 mg/L
- TDS resolution:0,1 / 1 / 10 / 100 ppm
- Temperature accuracy:± 0,8 °C
- Weight:0,39 kg (without protective covering)
- Cat. No.:LOVI724200
- pH accuracy:±0,02 + 2 digits
- pH resolution:0,01
- Data logging - log function:Automatic or manual
- Model:SensoDirect 150
- Conductivity range:0 - 200,0 µS / 0,2 - 2,000 mS / 2 - 20,00 mS / 20 - 200,00 mS
- Outputs:RS232
- TDS range:0 - 132 ppm / 132 - 1320 ppm / 1320 - 13200 ppm / 13200 - 132000 ppm
- mV accuracy:±0,5 % + 2 digits
- DO saturation:0 - 100,0 %
- Display:Large LCD with contrast adjustment
Accessories
- Description
- Description
- Description
- Description
- Description
- Description
- Description
Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.
Anders ontvangt u algemene aanbevelingen.



