Orders LinkContactUsLinkComponent
Comparator lighting units
Comparator lighting units
Daylight 2000 unit for Nessleriser, mains-operated
Catalog # 705-0682
Supplier:   Lovibond Tintometer
Comparator lighting units
Catalog # 705-0682
Comparator lighting units
Comparator lighting units
Daylight 2000 unit for Nessleriser, mains-operated
Price
1 010,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
For consistent and accurate readings when using Lovibond comparator instruments, it is important to use a constant light source for matching the sample with the glasses.
Standardised lighting unit, Daylight 2000: This mains-operated benchtop unit is corrected to good Northern daylight using a tungsten halogen lamp and a calibrated colour temperature-correction filter. It guarantees uniform lighting conditions for colour matching, allowing consistent and accurate readings when using Lovibond test discs.
Portable lighting units: The portable lighting unit is a constant light source powered by standard batteries for colour matching in any location.
Specifications
  • Description:
    Daylight 2000 unit for Nessleriser, mains-operated