Orders LinkContactUsLinkComponent
Labels, biohazard
Labels, biohazard
Biohazard labels, STN 104 -210X297-B7527, PP, Black on yellow, 210×297 mm, 1 label/card
Catalog # BRDY800094
Supplier:   Brady
Labels, biohazard
Catalog # BRDY800094
Supplier:  Brady
Labels, biohazard
Labels, biohazard
Biohazard labels, STN 104 -210X297-B7527, PP, Black on yellow, 210×297 mm, 1 label/card
Price
16,90 €
Log in for availability
1 * 1 ST
Product Details & Documents
All signs comply with current European legislation.
Ordering information: The polyester labels have a permanent acrylic adhesive.
Specifications
  • Colour:
    Black on yellow
  • Description:
    Biohazard labels
  • Dimensions:
    210×297 mm
  • Material:
    PP
  • Seq:
    9
  • Model:
    STN 104 -210X297-B7527
  • Packed:
    1 label/card