Blanking nuts
fit port size 1½" NPT
Catalog # COWIPC702.150
Supplier: COWIE
Jump to:
Product Details & Documents
PTFE, white.
- Chemically inert
- Reduce contamination and simplify cleaning
- Non-scratch
NPT blanking nuts/plugs are used to seal unused ports. To assemble simply screw the NPT blanking nut into a corresponding NPT port of the reactor lid.
Specifications
- Description:fit port size 1½" NPT
- Seq:05