Calibration weight sets, class E2, Proof Line
1 g - 1 kg, With DAkkS certificate
SARBYCS31-612-AC02
These class E2 weight sets include aluminium (1 to 5 mg) or nickel silver (10 to 500 mg) or stainless steel (1 g and higher) weights.
- Weight set:1 g - 1 kg
- Seq:09
- Type:With DAkkS certificate